siRNA Details
| Gene Symbol: DTWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173666.3 | NP_775937.1 | 1 | 107 | ||
| XM_011543338.2 | XP_011541640.2 | 1 | 1440 | |||
| GenBank mRNA | AK091296.1 | - | 1 | 47 | ||
| BC114508.1 | - | 1 | 43 | 40024023 | ||
| BC114955.1 | - | 1 | 43 | 40024022 | ||

DTWD2
Genomic Map