siRNA Details
| Gene Symbol: LINC00326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026969.1 | 4 | 868 | |||
| GenBank mRNA | BC039371.1 | - | 6 | 2170 | ||
| BC101702.1 | - | 2 | 243 | |||
| BC101704.1 | - | 2 | 242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00326| Gene Symbol: LINC00326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026969.1 | 4 | 868 | |||
| GenBank mRNA | BC039371.1 | - | 6 | 2170 | ||
| BC101702.1 | - | 2 | 243 | |||
| BC101704.1 | - | 2 | 242 | |||