siRNA Details
| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026873.1 | 5 | 2935 | |||
| GenBank mRNA | AK091213.1 | - | 1 | 1593 | ||
| AK092625.1 | - | 1 | 712 | |||
| AK124597.1 | - | 1 | 2056 | |||
| BC020399.1 | - | 5 | 2937 | |||
| BC093655.1 | - | 1 | 72 | |||
| BC093657.1 | - | 1 | 72 | |||

LINC00174
Genomic Map