siRNA Details
Gene Symbol: WEE2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015392.1 | 3 | 251 | |||
GenBank mRNA | AK098171.1 | - | 3 | 251 | ||
BC101358.2 | - | 2 | 56 | |||
BC131564.1 | - | 2 | 56 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WEE2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015392.1 | 3 | 251 | |||
GenBank mRNA | AK098171.1 | - | 3 | 251 | ||
BC101358.2 | - | 2 | 56 | |||
BC131564.1 | - | 2 | 56 |