siRNA Details
Gene Symbol: LINC00996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034033.1 | 2 | 717 | |||
GenBank mRNA | AK097158.1 | - | 2 | 619 | ||
BC109387.1 | - | 2 | 563 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034033.1 | 2 | 717 | |||
GenBank mRNA | AK097158.1 | - | 2 | 619 | ||
BC109387.1 | - | 2 | 563 |