siRNA Details
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 6 | 620 | |||
NR_024362.1 | 6 | 717 | ||||
GenBank mRNA | AK304567.1 | - | 6 | 689 | ||
BC035792.1 | - | 6 | 595 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 6 | 620 | |||
NR_024362.1 | 6 | 717 | ||||
GenBank mRNA | AK304567.1 | - | 6 | 689 | ||
BC035792.1 | - | 6 | 595 |