siRNA Details
| Gene Symbol: LINC01591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026706.2 | 1 | 221 | |||
| NR_120395.1 | 1 | 221 | ||||
| NR_120396.1 | 1 | 221 | ||||
| GenBank mRNA | BC045817.1 | - | 1 | 221 | ||
| BC117350.1 | - | 1 | 114 | |||
| BC117352.1 | - | 1 | 115 | |||
| BC144006.1 | - | 1 | 115 | |||
| BC171736.1 | - | 1 | 115 | |||

LINC01591
Genomic Map