siRNA Details
Gene Symbol: SLC25A25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033374.1 | 2, 3 | 933 | |||
GenBank mRNA | AK124723.1 | - | 2, 3 | 931 | ||
AL833509.1 | - | 2, 3 | 892 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC25A25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033374.1 | 2, 3 | 933 | |||
GenBank mRNA | AK124723.1 | - | 2, 3 | 931 | ||
AL833509.1 | - | 2, 3 | 892 |