siRNA Details
| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724640.2 | XP_006724703.1 | 1 | 42 | ||
| XM_011530922.2 | XP_011529224.1 | 1 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2BFM| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724640.2 | XP_006724703.1 | 1 | 42 | ||
| XM_011530922.2 | XP_011529224.1 | 1 | 205 | |||