siRNA Details
| Gene Symbol: LINC01203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045260.1 | 6 | 355 | |||
| GenBank mRNA | AA412296.1 | - | 2 | 224 | ||
| AA470018.1 | - | 6 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01203| Gene Symbol: LINC01203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045260.1 | 6 | 355 | |||
| GenBank mRNA | AA412296.1 | - | 2 | 224 | ||
| AA470018.1 | - | 6 | 355 | |||