siRNA Details
Gene Symbol: CSN1S2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003720.1 | 5, 6 | 198 | |||
GenBank mRNA | AY154892.1 | - | 5, 6 | 198 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CSN1S2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003720.1 | 5, 6 | 198 | |||
GenBank mRNA | AY154892.1 | - | 5, 6 | 198 |