siRNA Details
| Gene Symbol: EIF4E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134649.2 | NP_001128121.1 | 5, 6 | 542 | ||
| NM_001134650.1 | NP_001128122.1 | 5, 6 | 418 | |||
| NM_001134651.1 | NP_001128123.1 | 4, 5 | 409 | |||
| NM_001282886.1 | NP_001269815.1 | 5, 6 | 535 | |||
| NM_173359.4 | NP_775495.1 | 5, 6 | 503 | |||
| XM_017006278.1 | XP_016861767.1 | 6, 7 | 549 | |||
| XM_017006279.1 | XP_016861768.1 | 6, 7 | 536 | |||
| XM_017006280.1 | XP_016861769.1 | 6, 7 | 683 | |||
| XM_017006281.1 | XP_016861770.1 | 4, 5 | 411 | |||
| XM_017006282.1 | XP_016861771.1 | 6, 7 | 491 | |||
| GenBank mRNA | AK126999.1 | - | 4, 5 | 291 | ||
| AK313656.1 | - | 5, 6 | 503 | |||
| BC031289.2 | - | 3, 4 | 226 | 5299499 | ||
| BC068443.1 | - | 5, 6 | 535 | 4820931 | ||
| BX648727.1 | - | 1, 2 | 23 | |||
| DA227887.1 | - | 5, 6 | 393 | |||
| DA394405.1 | - | 5, 6 | 418 | |||
| DN990553.1 | - | 5, 6 | 550 | |||
| DR159502.1 | - | 4, 5 | 408 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025829, BC027014, AK005054, AY628329.

EIF4E3
Genomic Map