siRNA Details
| Gene Symbol: TTC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310135.1 | NP_001297064.1 | 13 | 3464 | ||
| XM_011537430.2 | XP_011535732.1 | 8 | 1272 | |||
| XM_011537431.2 | XP_011535733.1 | 6 | 929 | |||
| XM_011537432.2 | XP_011535734.1 | 13 | 3464 | |||
| XM_017021254.1 | XP_016876743.1 | 13 | 3464 | |||
| XM_017021255.1 | XP_016876744.1 | 13 | 3464 | |||
| XM_017021256.1 | XP_016876745.1 | 11 | 3554 | |||
| XM_017021257.1 | XP_016876746.1 | 13 | 3464 | |||

TTC6
Genomic Map