siRNA Details
| Gene Symbol: RAB37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006638.2 | NP_001006639.1 | 9 | 669 | ||
| NM_001163989.1 | NP_001157461.1 | 9 | 818 | |||
| NM_001163990.1 | NP_001157462.1 | 8 | 558 | |||
| NM_175738.4 | NP_783865.1 | 9 | 1062 | |||
| XM_005257288.4 | XP_005257345.1 | 9 | 687 | |||
| GenBank mRNA | AK054846.1 | - | 6 | 1797 | ||
| AK057069.1 | - | 3 | 213 | |||
| AK098068.1 | - | 9 | 669 | |||
| AK127073.1 | - | 8 | 1822 | |||
| AK290202.1 | - | 9 | 1038 | |||
| AK296062.1 | - | 9 | 657 | |||
| AK296172.1 | - | 8 | 521 | |||
| AK303442.1 | - | 9 | 721 | |||
| BC016615.1 | - | 9 | 615 | 4520191 | ||
| BC040547.1 | - | 9 | 1048 | 5287038 | ||

RAB37
Genomic Map