siRNA Details
| Gene Symbol: KRTAP8-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175857.3 | NP_787053.1 | 1 | 163 | ||
| GenBank mRNA | AB096961.1 | - | 1 | 161 | ||
| AJ457064.1 | - | 1 | 163 | |||
| BC130397.1 | - | 1 | 154 | 40146428 | ||
| BC133033.1 | - | 1 | 154 | 40147055 | ||

KRTAP8-1
Genomic Map