siRNA Details
| Gene Symbol: KRTAP19-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181607.1 | NP_853638.1 | 1 | 150 | ||
| GenBank mRNA | AB096943.1 | - | 1 | 182 | ||
| AJ457067.1 | - | 1 | 146 | |||
| BC126284.1 | - | 1 | 211 | 8992000 | ||
| BC126286.1 | - | 1 | 211 | 8992002 | ||

KRTAP19-1
Genomic Map