siRNA Details
| Gene Symbol: KRTAP13-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181621.3 | NP_853652.1 | 1 | 286 | ||
| GenBank mRNA | AB096938.1 | - | 1 | 276 | ||
| BC093934.1 | - | 1 | 285 | 7939779 | ||
| BC093936.1 | - | 1 | 286 | 7939781 | ||
| BX104478.1 | - | 1 | 303 | |||

KRTAP13-2
Genomic Map