siRNA Details
Gene Symbol: KRTAP19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181609.3 | NP_853640.1 | 1 | 327 | ||
GenBank mRNA | AF139541.1 | - | 1 | 327 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181609.3 | NP_853640.1 | 1 | 327 | ||
GenBank mRNA | AF139541.1 | - | 1 | 327 |