siRNA Details
| Gene Symbol: KRTAP20-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181616.1 | NP_853647.1 | 1 | 123 | ||
| GenBank mRNA | AB096957.1 | - | 1 | 151 | ||
| BC111084.1 | - | 1 | 123 | 40013739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP20-2| Gene Symbol: KRTAP20-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181616.1 | NP_853647.1 | 1 | 123 | ||
| GenBank mRNA | AB096957.1 | - | 1 | 151 | ||
| BC111084.1 | - | 1 | 123 | 40013739 | ||