siRNA Details
| Gene Symbol: ANKRD26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015928.1 | XP_016871417.1 | 39 | 7022 | ||
| XM_017015929.1 | XP_016871418.1 | 40 | 7010 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD26| Gene Symbol: ANKRD26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015928.1 | XP_016871417.1 | 39 | 7022 | ||
| XM_017015929.1 | XP_016871418.1 | 40 | 7010 | |||