siRNA Details
| Gene Symbol: LUZP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009909.3 | NP_001009909.2 | 5, 6 | 662 | ||
| NM_001252008.1 | NP_001238937.1 | 5, 6 | 658 | |||
| XM_017017648.1 | XP_016873137.1 | 5, 6 | 575 | |||
| GenBank mRNA | AL832641.3 | - | 5, 6 | 574 | ||
| BC132829.1 | - | 5, 6 | 454 | 40146851 | ||
| BC144177.1 | - | 5, 6 | 450 | |||

LUZP2
Genomic Map