siRNA Details
| Gene Symbol: WHAMMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003521.1 | 11 | 2545 | |||
| GenBank mRNA | BC048987.1 | - | 11 | 2605 | ||
| DQ309040.1 | - | 11 | 2545 | |||
| Gene Symbol: WHAMMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026589.1 | 9 | 4193 | |||
| GenBank mRNA | AK091254.1 | - | 4 | 2876 | ||

WHAMMP3
Genomic Map