siRNA Details
| Gene Symbol: ARL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040025.1 | NP_001035114.1 | 3 | 374 | ||
| GenBank mRNA | AK095847.1 | - | 2 | 640 | ||
| AK303526.1 | - | 3 | 203 | |||
| AK304718.1 | - | 3 | 213 | |||
| BC042380.1 | - | 3 | 212 | |||
| BC063687.1 | - | 3 | 176 | |||
| BC105076.1 | - | 3 | 374 | 8144079 | ||
| BC105078.1 | - | 3 | 374 | 8144081 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_197995, AK038349, BC139260, BF472168, BC139262, and others.

ARL16
Genomic Map