siRNA Details
| Gene Symbol: LOC101927481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126336.1 | 2 | 135 | |||
| GenBank mRNA | AV726231.1 | - | 2 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927481| Gene Symbol: LOC101927481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126336.1 | 2 | 135 | |||
| GenBank mRNA | AV726231.1 | - | 2 | 135 | ||