siRNA Details
| Gene Symbol: MYPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012643.3 | NP_001012661.1 | 2 | 269 | ||
| XM_017026742.1 | XP_016882231.1 | 1 | 1014 | |||
| XM_017026743.1 | XP_016882232.1 | 1 | 1014 | |||
| GenBank mRNA | BC044311.1 | - | 2 | 269 | 5756168 | |
| BU196283.1 | - | 161 | ||||

MYPOP
Genomic Map