siRNA Details
| Gene Symbol: KIF26B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018012.3 | NP_060482.2 | 3 | 1342 | ||
| GenBank mRNA | AK125187.1 | - | 3 | 1343 | ||
| AK293964.1 | - | 3 | 1257 | |||
| AK297303.1 | - | 3 | 1338 | |||
| AY923834.1 | - | 3 | 1343 | |||
| BC042481.1 | - | 2 | 676 | |||

KIF26B
Genomic Map