siRNA Details
| Gene Symbol: LOC339539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104171.1 | 4 | 1173 | |||
| GenBank mRNA | BC043541.1 | - | 4 | 1173 | ||
| BC105698.1 | - | 1 | 688 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC339539| Gene Symbol: LOC339539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104171.1 | 4 | 1173 | |||
| GenBank mRNA | BC043541.1 | - | 4 | 1173 | ||
| BC105698.1 | - | 1 | 688 | |||