siRNA Details
Gene Symbol: LINC00880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034007.1 | 4 | 1483 | |||
GenBank mRNA | BC040669.1 | - | 4 | 1478 | ||
BC122546.1 | - | 1 | 1083 |
Gene Symbol: LOC105374177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135546.1 | 1 | 356 | |||
GenBank mRNA | BC038760.1 | - | 1 | 355 |
