siRNA Details
| Gene Symbol: LINC00880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034007.1 | 4 | 1672 | |||
| GenBank mRNA | BC040669.1 | - | 4 | 1667 | ||
| BC122546.1 | - | 1 | 1272 | |||
| Gene Symbol: LOC105374177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135546.1 | 1 | 167 | |||
| GenBank mRNA | BC038760.1 | - | 1 | 167 | ||

LINC00880
Genomic Map