siRNA Details
| Gene Symbol: LRRC66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024611.2 | NP_001019782.1 | 4 | 2321 | ||
| XM_011534383.2 | XP_011532685.1 | 5 | 4042 | |||
| XM_017008162.1 | XP_016863651.1 | 3 | 5433 | |||
| GenBank mRNA | CR749811.1 | - | 4 | 2321 | ||

LRRC66
Genomic Map