siRNA Details
Gene Symbol: LOC101060498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121640.1 | 3 | 783 | |||
NR_121641.1 | 4 | 639 | ||||
GenBank mRNA | BM052690.1 | - | 1 | 183 | ||
JN120858.1 | - | 4 | 640 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101060498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121640.1 | 3 | 783 | |||
NR_121641.1 | 4 | 639 | ||||
GenBank mRNA | BM052690.1 | - | 1 | 183 | ||
JN120858.1 | - | 4 | 640 |