siRNA Details
Gene Symbol: LOC340107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037896.1 | 3 | 1743 | |||
GenBank mRNA | BC040315.1 | - | 3 | 1743 | ||
BC122542.1 | - | 1 | 207 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC340107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037896.1 | 3 | 1743 | |||
GenBank mRNA | BC040315.1 | - | 3 | 1743 | ||
BC122542.1 | - | 1 | 207 |