siRNA Details
| Gene Symbol: LOC340107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037896.1 | 3 | 1801 | |||
| GenBank mRNA | BC040315.1 | - | 3 | 1801 | ||
| BC122542.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC340107| Gene Symbol: LOC340107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037896.1 | 3 | 1801 | |||
| GenBank mRNA | BC040315.1 | - | 3 | 1801 | ||
| BC122542.1 | - | 1 | 265 | |||