siRNA Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207360.2 | NP_997243.2 | 2 | 537 | ||
| GenBank mRNA | AB458221.1 | - | 2 | 515 | ||
| AB458222.1 | - | 2 | 515 | |||
| AK127932.1 | - | 2 | 419 | |||
| BC127762.1 | - | 1 | 320 | 40127431 | ||
| BC157832.1 | - | 2 | 537 | 9057051 | ||

ZC3H12D
Genomic Map