siRNA Details
| Gene Symbol: KLHL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081675.2 | NP_001075144.2 | 1 | 690 | ||
| XM_005250901.4 | XP_005250958.1 | 2 | 911 | |||
| GenBank mRNA | AB196636.1 | - | 1 | 666 | ||
| BC127886.1 | - | 1 | 690 | 40131758 | ||

KLHL38
Genomic Map