siRNA Details
| Gene Symbol: LINC01492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121578.1 | 6, 7 | 782 | |||
| GenBank mRNA | BC035187.1 | - | 6, 7 | 787 | ||
| BU852916.1 | - | 2, 3 | 304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01492| Gene Symbol: LINC01492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121578.1 | 6, 7 | 782 | |||
| GenBank mRNA | BC035187.1 | - | 6, 7 | 787 | ||
| BU852916.1 | - | 2, 3 | 304 | |||