siRNA Details
| Gene Symbol: LRIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017924.3 | NP_001017924.1 | 3 | 1223 | ||
| NM_001284223.1 | NP_001271152.1 | 4 | 1253 | |||
| GenBank mRNA | BC140765.1 | - | 4 | 1253 | 9021633 | |
| BC144475.1 | - | 4 | 1253 | 9053002 | ||
| BX648524.1 | - | 3 | 1163 | |||

LRIT2
Genomic Map