siRNA Details
| Gene Symbol: C12orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080406.1 | NP_001073875.1 | 1, 2 | 548 | ||
| XM_011520847.2 | XP_011519149.1 | 2, 3 | 1090 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf71| Gene Symbol: C12orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080406.1 | NP_001073875.1 | 1, 2 | 548 | ||
| XM_011520847.2 | XP_011519149.1 | 2, 3 | 1090 | |||