siRNA Details
| Gene Symbol: SLC35F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101318.1 | - | 11 | |||
| BC101319.1 | - | 1 | 17 | |||
| BC101320.1 | - | 1 | 17 | |||
| BC101321.1 | - | 1 | 17 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC35F4| Gene Symbol: SLC35F4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101318.1 | - | 11 | |||
| BC101319.1 | - | 1 | 17 | |||
| BC101320.1 | - | 1 | 17 | |||
| BC101321.1 | - | 1 | 17 | |||