siRNA Details
Gene Symbol: FMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103184.3 | NP_001096654.1 | 1 | 1026 | ||
GenBank mRNA | AL833157.1 | - | 1 | 1157 | ||
BC103692.1 | - | 1 | 1026 | |||
BC107593.1 | - | 1 | 1026 |
