siRNA Details
| Gene Symbol: ZKSCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012981.4 | NP_001012999.3 | 6 | 2012 | ||
| XM_017023200.1 | XP_016878689.1 | 6 | 2432 | |||
| GenBank mRNA | AK302399.1 | - | 6 | 2048 | ||
| AK303472.1 | - | 6 | 2409 | |||
| BC130007.1 | - | 6 | 1670 | 40132986 | ||
| BC151139.1 | - | 6 | 1656 | 9057039 | ||
| BX648785.1 | - | 6 | 2000 | |||

ZKSCAN2
Genomic Map