siRNA Details
| Gene Symbol: ZNF404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033719.2 | NP_001028891.2 | 2 | 1281 | ||
| GenBank mRNA | AK300120.1 | - | 2 | 1325 | ||
| BC101332.1 | - | 2 | 1275 | |||
| BC101333.1 | - | 2 | 1281 | 40025019 | ||
| BQ188692.1 | - | 1 | 17 | |||

ZNF404
Genomic Map