siRNA Details
| Gene Symbol: ZNF284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037813.2 | NP_001032902.1 | 5 | 560 | ||
| XM_011526907.2 | XP_011525209.1 | 5 | 601 | |||
| XM_011526908.2 | XP_011525210.1 | 6 | 661 | |||
| GenBank mRNA | AK308295.1 | - | 11 | 2269 | ||
| AY166789.1 | - | 3 | 262 | |||
| AY458845.1 | - | 5 | 560 | |||
| CR936662.1 | - | 5 | 503 | |||

ZNF284
Genomic Map