siRNA Details
| Gene Symbol: LOC105371448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003084.1 | XP_016858573.1 | 2 | 769 | ||
| XM_017003085.1 | XP_016858574.1 | 2 | 769 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371448| Gene Symbol: LOC105371448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003084.1 | XP_016858573.1 | 2 | 769 | ||
| XM_017003085.1 | XP_016858574.1 | 2 | 769 | |||