siRNA Details
| Gene Symbol: OR2T8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005522.1 | NP_001005522.1 | 1 | 287 | ||
| XM_011544178.2 | XP_011542480.1 | 2 | 1630 | |||
| XM_017001172.1 | XP_016856661.1 | 2 | 4409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2T8| Gene Symbol: OR2T8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005522.1 | NP_001005522.1 | 1 | 287 | ||
| XM_011544178.2 | XP_011542480.1 | 2 | 1630 | |||
| XM_017001172.1 | XP_016856661.1 | 2 | 4409 | |||