siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 6 | 956 | |||
| GenBank mRNA | AK310337.1 | - | 6 | 956 | ||
| DV080387.1 | - | 1 | 44 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf173| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 6 | 956 | |||
| GenBank mRNA | AK310337.1 | - | 6 | 956 | ||
| DV080387.1 | - | 1 | 44 | |||