siRNA Details
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026933.2 | 6 | 1094 | |||
GenBank mRNA | DV080387.1 | - | 1 | 182 | ||
DW009696.1 | - | 5 | 523 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026933.2 | 6 | 1094 | |||
GenBank mRNA | DV080387.1 | - | 1 | 182 | ||
DW009696.1 | - | 5 | 523 |