siRNA Details
| Gene Symbol: OSTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198184.1 | NP_937827.1 | 2 | 296 | ||
| XM_005247428.4 | XP_005247485.1 | 3 | 544 | |||
| XM_017006303.1 | XP_016861792.1 | 3 | 455 | |||
| GenBank mRNA | AY398681.1 | - | 2 | 296 | ||
| AY573933.1 | - | 2 | 296 | |||
| BC128106.1 | - | 2 | 296 | 40112103 | ||
| BC128107.1 | - | 2 | 296 | 40112104 | ||

OSTN
Genomic Map