siRNA Details
| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198505.2 | NP_940907.2 | 25 | 2831 | ||
| XM_011512770.1 | XP_011511072.1 | 25 | 2930 | |||
| XM_017006304.1 | XP_016861793.1 | 21 | 2144 | |||
| XM_017006305.1 | XP_016861794.1 | 12 | 1247 | |||
| GenBank mRNA | AY823163.1 | - | 25 | 2831 | ||

ATP13A5
Genomic Map