siRNA Details
| Gene Symbol: MSANTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042690.1 | NP_001036155.1 | 2 | 606 | ||
| XM_011513465.2 | XP_011511767.1 | 5 | 1868 | |||
| XM_011513466.2 | XP_011511768.1 | 3 | 4133 | |||
| XM_017008169.1 | XP_016863658.1 | 3 | 3311 | |||
| GenBank mRNA | AK126094.1 | - | 2 | 606 | ||
| BC043530.1 | - | 5 | 1227 | |||

MSANTD1
Genomic Map